4-Chlorophenylboronic acid, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
417548-10G 417548-1G 417548-25G
SIAL417548-10GEA 104 EUR
SIAL417548-10G SIAL417548-1G SIAL417548-25G
4-Chlorophenylboronic acid, Sigma-Aldrich®
4-Chlorophenylboronic acid
Formula: C₆H₆BClO₂
Melting Pt: 268…272 °C
Density: ∼1 g/cm³ (20 °C)
MDL Number: MFCD00039137
CAS Number: 1679-18-1
EINECS: 216-845-5