2,6-Dichlorobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
D56500-100G D56500-25G
SIALD56500-100GEA 45.5 EUR
SIALD56500-100G SIALD56500-25G
2,6-Dichlorobenzaldehyde, Sigma-Aldrich®
2,6-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 239
Melting Pt: 69…70 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003307
CAS Number: 83-38-5
EINECS: 201-472-2