3-Chlorobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
C23403-100G C23403-25G
SIALC23403-100GEA 109 EUR
SIALC23403-100G SIALC23403-25G
3-Chlorobenzaldehyde, Sigma-Aldrich®
3-Chlorobenzaldehyde
Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 214
Melting Pt: 17…18 °C
Density: 1.241 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003350
CAS Number: 587-04-2
EINECS: 209-596-9