4-Chlorobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
112216-250G 112216-50G
SIAL112216-250GEA 58.9 EUR
SIAL112216-250G SIAL112216-50G
4-Chlorobenzaldehyde, Sigma-Aldrich®
4-Chlorobenzaldehyde
Formula: C₇H₅ClO
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: 46 °C
Density: 1.196 g/cm³ (20 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00003379
CAS Number: 104-88-1
EINECS: 203-247-4
UN: 3077
ADR: 9,III