3,4-Dichlorobenzoyl chloride ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
A17006.14
A17006.22
A17006.14EA
55.8
EUR
A17006.14
A17006.22
3,4-Dichlorobenzoyl chloride ≥97%
3,4-Dichlorobenzoyl chloride
Purity: 97%
CAS No.: 3024-72-4
Molecular Formula: C7H3Cl3O
Formula Weight: 209.46
Flash Point: 142°(287°F)
Melting Point: 30-34°
Boiling Point: 242°
Beilstein Reference: 607485
UN No.: 3261
MDL No.: MFCD00000672
Formula:
C₇H₃Cl₃O Boiling Pt: 242 °C Melting Pt: 30…34 °C Flash Pt: 142 °C (287 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000672 CAS Number: 3024-72-4 EINECS: 221-177-2 UN: 3261 ADR: 8,II |