4-Chlorobenzoyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: p-Chlorobenzoyl chloride
M-Clarity™ quality level = MQ200
111902-5G 111902-100G 111902-500G
SIAL111902-5GEA 38.3 EUR
SIAL111902-5G SIAL111902-100G SIAL111902-500G
4-Chlorobenzoyl chloride, Sigma-Aldrich®
4-Chlorobenzoyl chloride
Formula: C₇H₄Cl₂O
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 14…15 °C
Density: 1.377 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000686
CAS Number: 122-01-0
EINECS: 204-515-3
UN: 3265
ADR: 8,II