2,5-Dichlorobenzoic acid, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
144940-250G 144940-50G
SIAL144940-250GEA 232 EUR
SIAL144940-250G SIAL144940-50G
2,5-Dichlorobenzoic acid, Sigma-Aldrich®
2,5-Dichlorobenzoic acid
Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Boiling Pt: 301
Melting Pt: 151…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00002416
CAS Number: 50-79-3
EINECS: 200-065-7