1-Chloro-4-iodobenzene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
101605-100G
101605-25G
SIAL101605-100GEA
92
EUR
SIAL101605-100G
SIAL101605-25G
1-Chloro-4-iodobenzene, Sigma-Aldrich®
1-Chloro-4-iodobenzene
Formula:
C₆H₄ClI Boiling Pt: 226 °C (1013 hPa) Melting Pt: 57 °C Density: 1.886 g/cm³ (57 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001053 CAS Number: 637-87-6 EINECS: 211-305-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...