4-Chlorophenyl isocyanate, Sigma-Aldrich®
Supplier: Merck
152277-100G 152277-5G
SIAL152277-100GEA 109 EUR
SIAL152277-100G SIAL152277-5G
4-Chlorophenyl isocyanate, Sigma-Aldrich®
4-Chlorophenyl isocyanate
Formula: C₇H₄ClNO
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 31…32 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002024
CAS Number: 104-12-1
EINECS: 203-176-9
UN: 2811
ADR: 6.1,I