3,5-Dichloroaniline, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
D55792-100G D55792-10G
SIALD55792-100GEA 60.2 EUR
SIALD55792-100G SIALD55792-10G
3,5-Dichloroaniline, Sigma-Aldrich®
3,5-Dichloroaniline
Formula: C₆H₅Cl₂N
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 51 °C
Density: 1.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007774
CAS Number: 626-43-7
EINECS: 210-948-9
UN: 3442
ADR: 6.1,II