1-Chloro-2-nitrobenzene, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1-Chloro-2-nitrobenzene, 2-Chloronitrobenzene
M-Clarity™ quality level = MQ200
185760-1KG
185760-250G
SIAL185760-1KGEA
78.8
EUR
SIAL185760-1KG
SIAL185760-250G
1-Chloro-2-nitrobenzene, Sigma-Aldrich®
1-Chloro-2-nitrobenzene
Formula:
C₆H₄ClNO₂ Boiling Pt: 246 °C (1013 hPa) Melting Pt: 33 °C Density: 1.368 g/cm³ (20 °C) Flash Pt: 128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007061 CAS Number: 88-73-3 EINECS: 201-854-9 UN: 1578 ADR: 6.1,II Merck Index: 13,02170 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...