2-Chloro-4-nitrophenol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Chloro-2-hydroxy-5-nitrobenzene
M-Clarity™ quality level = MQ100
C61208-10G C61208-50G
SIALC61208-10GEA 35.4 EUR
SIALC61208-10G SIALC61208-50G
2-Chloro-4-nitrophenol, Sigma-Aldrich®
2-Chloro-4-nitrophenol
Formula: C₆H₄ClNO₃
MW: 173.56 g/mol
Melting Pt: 109…112 °C
MDL Number: MFCD00043910
CAS Number: 619-08-9
EINECS: 210-578-8