Benzonitrile, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Cyanobenzene, Phenylcyanide
M-Clarity™ quality level = MQ100
294098-100ML 294098-1L
SIAL294098-100MLEA 111 EUR
SIAL294098-100ML SIAL294098-1L
Benzonitrile, Sigma-Aldrich®
Benzonitrile
Formula: C₇H₅N
MW: 103.12 g/mol
Boiling Pt: 190.7
Melting Pt: –13 °C
Density: 1 g/cm³ (20 °C)
Flash Pt: 70
Storage Temperature: Ambient
MDL Number: MFCD00001770
CAS Number: 100-47-0
EINECS: 202-855-7
UN: 2224
ADR: 6.1 II
Merck Index: 13,01098