3-Cyanobenzoyl chloride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

302538-1G 302538-5G
SIAL302538-1GEA 75.5 EUR
SIAL302538-1G SIAL302538-5G
3-Cyanobenzoyl chloride, Sigma-Aldrich®
3-Cyanobenzoyl chloride
Formula: C₈H₄ClNO
MW: 165.58 g/mol
MDL Number: MFCD00011545
CAS Number: 1711-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR