4-Cyanobenzoyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Danger
B24333.06 B24333.14 B24333.22
B24333.06EA 59.2 EUR
B24333.06 B24333.14 B24333.22
4-Cyanobenzoyl chloride ≥98%
4-Cyanobenzoyl chloride
Purity: 98% CAS No.: 6068-72-0 Molecular Formula: C8H4ClNO Formula Weight: 165.58 Odour: Irritating Melting Point: 68-70° Beilstein Reference: 386729 UN No.: 3261 MDL No.: MFCD00001822
Formula: C₈H₄ClNO
MW: 165.58 g/mol
Boiling Pt: 150…151 °C (25 mmHg)
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00001822
CAS Number: 6068-72-0
EINECS: 228-005-5
UN: 3261
ADR: 8 II