4-Cyanobenzoyl chloride, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

124826-25G 124826-5G 124826-1G
SIAL124826-25GEA 143 EUR
SIAL124826-25G SIAL124826-5G SIAL124826-1G
4-Cyanobenzoyl chloride, Sigma-Aldrich®
4-Cyanobenzoyl chloride
Formula: C₈H₄ClNO
Boiling Pt: 150…151 °C (25 mmHg)
Melting Pt: 78.5 °C
MDL Number: MFCD00001822
CAS Number: 6068-72-0
EINECS: 228-005-5
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR