Anthranilonitrile, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Cyanoaniline, o-Cyanoaniline, 2-Aminobenzonitrile, o-Aminobenzonitrile
M-Clarity™ quality level = MQ200
A89901-100G A89901-25G
SIALA89901-100GEA 144 EUR
SIALA89901-100G SIALA89901-25G
Anthranilonitrile, Sigma-Aldrich®
Anthranilonitrile
Formula: C₇H₆N₂
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 47 °C
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007631
CAS Number: 1885-29-6
EINECS: 217-549-9
UN: 3439
ADR: 6.1,III