o-Acetoxybenzoyl chloride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
165190-25G
165190-5G
SIAL165190-25GEA
80
EUR
SIAL165190-25G
SIAL165190-5G
o-Acetoxybenzoyl chloride, Sigma-Aldrich®
o-Acetoxybenzoyl chloride
Formula:
C₉H₇ClO₃ Boiling Pt: 346 °C (1013 hPa) Melting Pt: 45…48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000663 CAS Number: 5538-51-2 EINECS: 226-899-1 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...