O-Acetylsalicylic acid, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
A3160-1VL
SIALA3160-1VLEA
140
EUR
SIALA3160-1VL
O-Acetylsalicylic acid, Sigma-Aldrich®
o-Acetylsalicylic acid
Formula:
2-(CH₃CO₂)C₆H₄CO₂H Boiling Pt: 140 °C (1013 hPa) Melting Pt: 138…140 °C Density: 1.39 g/cm³ (20 °C) Flash Pt: 250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002430 CAS Number: 50-78-2 EINECS: 200-064-1 Merck Index: 13,00856 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...