2-Nitrobenzoyl chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
322717-25G 322717-5G
SIAL322717-25GEA 122 EUR
SIAL322717-25G SIAL322717-5G
2-Nitrobenzoyl chloride, Sigma-Aldrich®
2-Nitrobenzoyl chloride
Formula: C₇H₄ClNO₃
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 25 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007131
CAS Number: 610-14-0
EINECS: 210-206-4
UN: 3261
ADR: 8,II