3-Nitrobenzoyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: m-Nitrobenzoyl chloride
A10579.0B A10579.18 A10579.30
A10579.0BEA 0 EUR
A10579.0B A10579.18 A10579.30
3-Nitrobenzoyl chloride ≥98%
3-Nitrobenzoyl chloride
Purity: 98% CAS No.: 121-90-4 Molecular Formula: C7H4ClNO3 Formula Weight: 185.57 Flash Point: 110°(230°F) Odour: Pungent Boiling Point: 276-278° dec. Beilstein Reference: 777186 UN No.: 3261 MDL No.: MFCD00007247
Formula: C₇H₄ClNO₃
Boiling Pt: 276…278 °C (dec.)
Melting Pt: 31…35 °C
Density: 1.42
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007247
CAS Number: 121-90-4
EINECS: 204-505-9
UN: 3261
ADR: 8,II