4-Hexyloxybenzoyl chloride ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
A13367.06
A13367.14
A13367.22
A13367.06EA
66.8
EUR
A13367.06
A13367.14
A13367.22
4-Hexyloxybenzoyl chloride ≥98%
4-Hexyloxybenzoyl chloride
Purity: 98%
CAS No.: 39649-71-3
Molecular Formula: C13H17ClO2
Formula Weight: 240.73
Flash Point:
Formula:
C₁₃H₁₇ClO₂ Boiling Pt: 203…205 °C (15 mmHg) Density: 1.082 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000690 CAS Number: 39649-71-3 EINECS: 254-561-3 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...