4-Anisoyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 4-Methoxybenzoyl chloride
M-Clarity™ quality level = MQ200
A88476-100G A88476-25G A88476-5G
SIALA88476-100GEA 127 EUR
SIALA88476-100G SIALA88476-25G SIALA88476-5G
4-Anisoyl chloride, Sigma-Aldrich®
4-Anisoyl chloride
Formula: C₈H₇ClO₂
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 82…83 °C
Density: 1.261 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000687
CAS Number: 100-07-2
EINECS: 202-816-4
UN: 1729
ADR: 8,II
Merck Index: 13,00676