Benzamide ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A10501.0E
A10501.22
A10501.36
A10501.0EEA
265
EUR
A10501.0E
A10501.22
A10501.36
Benzamide ≥98%
Benzamide
Purity: 98+%
CAS No.: 55-21-0
Molecular Formula: C7H7NO
Formula Weight: 121.14
Flash Point: 180°(356°F)
Melting Point: 125-129°
Boiling Point: 288°
Beilstein Reference: 385876
Merck Reference: 14,1060
MDL No.: MFCD00007968
Formula:
C₇H₇NO Boiling Pt: 288 °C Melting Pt: 125…129 °C Density: 1.341 Flash Pt: 180 °C (356 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007968 CAS Number: 55-21-0 EINECS: 200-227-7 Merck Index: 14,1060 |