Benzamide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Benzoic acid amide
M-Clarity™ quality level = MQ100
135828-5G 135828-100G
SIAL135828-5GEA 35.2 EUR
SIAL135828-5G SIAL135828-100G
Benzamide, Sigma-Aldrich®
Benzamide
Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 288
Melting Pt: 124
Density: 1.35 g/cm³ (20 °C)
Flash Pt: 180
Storage Temperature: Ambient
MDL Number: MFCD00007968
CAS Number: 55-21-0
EINECS: 200-227-7
Merck Index: 13,01059