Butyl benzoate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Benzoic acid butyl ester
M-Clarity™ quality level = MQ100
293296-1KG 293296-250G
SIAL293296-1KGEA 28.7 EUR
SIAL293296-1KG SIAL293296-250G
Butyl benzoate, Sigma-Aldrich®
Butyl benzoate
Formula: C₁₁H₁₄O₂
Boiling Pt: 249…251 °C (1013 hPa)
Melting Pt: –22 °C
Density: 1.01 g/cm³ (20 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00009439
CAS Number: 136-60-7
EINECS: 205-252-7
Merck Index: 13,01551