Ethyl benzoate, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Benzoic acid ethyl ester

M-Clarity™ quality level = MQ200

E12907-100G E12907-500G E12907-5G
SIALE12907-100GEA 28.1 EUR
SIALE12907-100G SIALE12907-500G SIALE12907-5G
Ethyl benzoate, Sigma-Aldrich®
Ethyl benzoate
Formula: C₉H₁₀O₂
Boiling Pt: 415 °C (1013 hPa)
Melting Pt: –35 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00009109
CAS Number: 93-89-0
EINECS: 202-284-3
Merck Index: 13,03802

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR