Benzoic acid methyl ester, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Methyl benzoate
M-Clarity™ quality level = MQ300
W268305-1KG W268305-25KG W268305-SAMPLE
SIALW268305-1KGEA 71.6 EUR
SIALW268305-1KG SIALW268305-25KG SIALW268305-SAMPLE
Benzoic acid methyl ester, Sigma-Aldrich®
Benzoic acid methyl ester
Formula: C₆H₅COOCH₃
MW: 136.15 g/mol
Boiling Pt: 199.6
Melting Pt: –12 °C
Density: 1.0888 g/cm³ (20 °C)
Flash Pt: 83
Storage Temperature: Ambient
MDL Number: MFCD00008421
CAS Number: 93-58-3
EINECS: 202-259-7
Merck Index: 13,06052
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=287e16081e44f5525ff8b4d34f0a6fece44a3073&vid=2be9af3b90e55af477c374c091cd0888dcf60b96&prid=web&clid=SAPDAM