Methyl-4-hydroxybenzoate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
M-Clarity™ quality level = MQ300
W271004-10KG-K W271004-1KG-K W271004-5KG-K
SIALW271004-10KG-KEA 713 EUR
SIALW271004-10KG-K SIALW271004-1KG-K SIALW271004-5KG-K
Methyl-4-hydroxybenzoate, Sigma-Aldrich®
Methyl 4-hydroxybenzoate
Formula: HOC₆H₄CO₂CH₃
MW: 152.15 g/mol
Boiling Pt: 270…280 °C (1013 hPa)
Melting Pt: 125…128 °C
Density: 1.1208 g/cm³ (137 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002352
CAS Number: 99-76-3
EINECS: 202-785-7
Merck Index: 13,06129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84321052052f4be2a53d8d7ebac7fabe6fc5a435&vid=ede765fad275b2515a8c57ba4499bff313a18983&prid=web&clid=SAPDAM