Diethyl phthalate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W512206-1KG W512206-25KG W512206-SAMPLE
SIALW512206-1KGEA 80.7 EUR
SIALW512206-1KG SIALW512206-25KG SIALW512206-SAMPLE
Diethyl phthalate, Sigma-Aldrich®
Diethyl phthalate
Formula: C₁₂H₁₄O₄
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: –3 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 156 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00009111
CAS Number: 84-66-2
EINECS: 201-550-6
Merck Index: 12,07527
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d0a6c6b14ead75148c0661141b5e8d00e6fc1a9&vid=2461e9f0e230ca44b5372d1233410fbf47a564c7&prid=web&clid=SAPDAM