4-(Dimethylamino)benzoic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Dimethylaminobenzoic acid
M-Clarity™ quality level = MQ200
D139459-100G D139459-25G
SIALD139459-100GEA 99.8 EUR
SIALD139459-100G SIALD139459-25G
4-(Dimethylamino)benzoic acid, Sigma-Aldrich®
4-(Dimethylamino)benzoic acid
Formula: C₉H₁₁NO₂
Boiling Pt: 297 °C (1013 hPa)
Melting Pt: 149 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002537
CAS Number: 619-84-1
EINECS: 210-615-8
Merck Index: 13,03259