5-Aminosalicylic acid
Supplier: Cayman Chemical
Synonyms: Mesalamine, Mesalazine, 5-Amino-2-hydroxybenzoic acid, m-Aminosalicylic acid
70265-10 70265-25 70265-50
CAYM70265-10EA 22 EUR
CAYM70265-10 CAYM70265-25 CAYM70265-50
5-Aminosalicylic acid
5-Aminosalicylic acid
Formula: C₇H₇NO₃
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 280 °C
Density: 1.568 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007877
CAS Number: 89-57-6
EINECS: 201-919-1
Merck Index: 12,05964