m-Anisic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Anisic acid, 3-Methoxybenzoic acid
MQ100
117714-100G 117714-25G
SIAL117714-100GEA 78.9 EUR
SIAL117714-100G SIAL117714-25G
m-Anisic acid, Sigma-Aldrich®
m-Anisic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 308
Melting Pt: 103…106 °C
Density: 1.14 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002499
CAS Number: 586-38-9
EINECS: 209-574-9