3-Hydroxybenzoic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: m-Hydroxybenzoic Acid
M-Clarity™ quality level = MQ200
H20008-100G H20008-500G H20008-5G
SIALH20008-100GEA 57.3 EUR
SIALH20008-100G SIALH20008-500G SIALH20008-5G
3-Hydroxybenzoic acid, Sigma-Aldrich®
3-Hydroxybenzoic acid
Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 346
Melting Pt: 199
Density: 1.473 g/cm³ (20 °C)
Flash Pt: 220
Storage Temperature: Ambient
MDL Number: MFCD00002506
CAS Number: 99-06-9
EINECS: 202-726-5