1,3-Difluorobenzene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
D102008-100G D102008-10G
SIALD102008-100GEA 136 EUR
SIALD102008-100G SIALD102008-10G
1,3-Difluorobenzene, Sigma-Aldrich®
1,3-Difluorobenzene
Formula: C₆H₄F₂
Boiling Pt: 83 °C (1013 hPa)
Melting Pt: –59 °C
Density: 1.16 g/cm³ (20 °C)
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00000327
CAS Number: 372-18-9
EINECS: 206-746-5
UN: 1993
ADR: 3,II