4-Fluorophenylboronic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: (4-Fluorophenyl)metaboric acid, 4-Fluorobenzeneboronic acid, Dihydroxy(4-fluorophenyl)borane
M-Clarity™ quality level = MQ100
417556-1G 417556-25G 417556-5G
SIAL417556-1GEA 31.1 EUR
SIAL417556-1G SIAL417556-25G SIAL417556-5G
4-Fluorophenylboronic acid, Sigma-Aldrich®
4-Fluorophenylboronic acid
Formula: C₆H₆BFO₂
Melting Pt: 263…265 °C
Storage Temperature: Ambient
MDL Number: MFCD00039136
CAS Number: 1765-93-1