3-(Trifluoromethoxy)phenylboronic acid 98%
Supplier: Thermo Scientific
Warning
Synonyms: (3-(Trifluoromethoxy)phenyl)metaboric acid, (3-(Trifluoromethoxy)phenyl)boronic acid, 3-(Trifluoromethoxy)benzeneboronic acid, Dihydroxy(3-(trifluoromethoxy)phenyl)borane
AC35972.250 AC35972.1
MAYBAC35972.250EA 0 EUR
MAYBAC35972.250 MAYBAC35972.1
3-(Trifluoromethoxy)phenylboronic acid 98%
3-(Trifluoromethoxy)phenylboronic acid
Formula: C₇H₆BF₃O₃
MW: 205.93 g/mol
Melting Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD01320697
CAS Number: 179113-90-7
Order Now
Specification Test Results
Appearance White to light beige Powder and/or chunks
Infrared spectrum Conforms
HPLC ≥97.5 %