3,4-Difluorobenzaldehyde, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
265160-25G 265160-5G
SIAL265160-25GEA 149 EUR
SIAL265160-25G SIAL265160-5G
3,4-Difluorobenzaldehyde, Sigma-Aldrich®
3,4-Difluorobenzaldehyde
Formula: C₇H₄F₂O
Boiling Pt: 53 °C (1013 hPa)
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010328
CAS Number: 34036-07-2
EINECS: 422-180-3