Pentafluorobenzaldehyde, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

103748-10G 103748-2.5G
SIAL103748-10GEA 111 EUR
SIAL103748-10G SIAL103748-2.5G
Pentafluorobenzaldehyde, Sigma-Aldrich®
Pentafluorobenzaldehyde
Formula: C₇HF₅O
Boiling Pt: 164 °C (1013 hPa)
Melting Pt: 24 °C
Density: 1.7518 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003303
CAS Number: 653-37-2
EINECS: 211-502-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR