3-(Trifluoromethoxy)benzonitrile

Supplier: Thermo Scientific

SB01895.1 SB01895.10
MAYBSB01895.1EA 0 EUR
MAYBSB01895.1 MAYBSB01895.10
3-(Trifluoromethoxy)benzonitrile
3-(Trifluoromethoxy)benzonitrile
Formula: C₈H₄F₃NO
Boiling Pt: 236 °C (1013 hPa)
Melting Pt: 37 °C
MDL Number: MFCD00040959
CAS Number: 52771-22-9
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR