2,3,4,5-Tetrafluorobenzoyl chloride
Supplier: Thermo Scientific
Danger
SB02060.1 SB02060.10 SB02060.50
MAYBSB02060.1EA 0 EUR
MAYBSB02060.1 MAYBSB02060.10 MAYBSB02060.50
2,3,4,5-Tetrafluorobenzoyl chloride
2,3,4,5-Tetrafluorobenzoyl chloride
Formula: C₇HClF₄O
Boiling Pt: 53…54 °C (1013 hPa)
Density: 1.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00075164
CAS Number: 94695-48-4
UN: 3265
ADR: 8,II