4-Fluorobenzoyl chloride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
p-Fluorobenzoyl chloride
M-Clarity™ quality level = MQ200
119946-100G
119946-25G
119946-5G
SIAL119946-100GEA
127
EUR
SIAL119946-100G
SIAL119946-25G
SIAL119946-5G
4-Fluorobenzoyl chloride, Sigma-Aldrich®
4-Fluorobenzoyl chloride
Formula:
C₇H₄ClFO Boiling Pt: 180 °C (1013 hPa) Melting Pt: 9 °C Density: 1.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000684 CAS Number: 403-43-0 EINECS: 206-961-4 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...