Methyl-2-fluorobenzoate ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A16133.06
A16133.14
A16133.06EA
14.8
EUR
A16133.06
A16133.14
Methyl-2-fluorobenzoate ≥98%
Methyl 2-fluorobenzoate
Purity: 98%
CAS No.: 394-35-4
Molecular Formula: C8H7FO2
Formula Weight: 154.14
Flash Point: 93°(199°F)
Refractive Index: 1.5030
Odour: Aromatic
Boiling Point: 109-110°/35mm
Beilstein Reference: 1862493
MDL No.: MFCD00017913
Formula:
C₈H₇FO₂ Boiling Pt: 109…110 °C (35 mmHg) Density: 1.210 Flash Pt: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00017913 CAS Number: 394-35-4 EINECS: 206-894-0 |