Methyl-2-fluorobenzoate

Supplier: Thermo Scientific

SPB03422.1 SPB03422.50
MAYBSPB03422.1EA 0 EUR
MAYBSPB03422.1 MAYBSPB03422.50
Methyl-2-fluorobenzoate
Methyl 2-fluorobenzoate
Formula: C₈H₇FO₂
MDL Number: MFCD00017913
CAS Number: 394-35-4
EINECS: 206-894-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR