4-Fluoro-3-nitrobenzoic acid, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

329045-25G 329045-5G
SIAL329045-25GEA 76.3 EUR
SIAL329045-25G SIAL329045-5G
4-Fluoro-3-nitrobenzoic acid, Sigma-Aldrich®
4-Fluoro-3-nitrobenzoic acid
Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Boiling Pt: 92 °C (15 mmHg)
Melting Pt: 123…124 °C
MDL Number: MFCD00007058
CAS Number: 453-71-4
EINECS: 207-221-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR