5-Fluorosalicylic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: 5-Fluoro-2-hydroxybenzoic acid
M-Clarity™ quality level = MQ100
152706-1G 152706-5G
SIAL152706-1GEA 29.3 EUR
SIAL152706-1G SIAL152706-5G
5-Fluorosalicylic acid, Sigma-Aldrich®
5-Fluorosalicylic acid
Formula: C₇H₅FO₃
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 180 °C
MDL Number: MFCD00002456
CAS Number: 345-16-4
EINECS: 206-455-3