2-Fluorophenyl isocyanate, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Fluoro-2-isocyanatobenzene

M-Clarity™ quality level = MQ100

159352-25G 159352-5G
SIAL159352-25GEA 81.7 EUR
SIAL159352-25G SIAL159352-5G
2-Fluorophenyl isocyanate, Sigma-Aldrich®
2-Fluorophenyl isocyanate
Formula: C₇H₄FNO
Boiling Pt: 41…44 °C (1 mmHg)
Density: 1.222 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001996
CAS Number: 16744-98-2
EINECS: 240-804-0
UN: 3080
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR