4-(Trifluoromethoxy)phenyl isocyanate

Supplier: Thermo Scientific

Synonyms: 1-Isocyanato-4-(trifluoromethoxy)benzene

SB01053DA SB01053EA
MAYBSB01053DAEA 0 EUR
MAYBSB01053DA MAYBSB01053EA
4-(Trifluoromethoxy)phenyl isocyanate
4-(Trifluoromethoxy)phenyl isocyanate
Formula: C₈H₄F₃NO₂
Boiling Pt: 78 °C (22 mmHg)
Density: 1.34 g/cm³ (20 °C)
MDL Number: MFCD00035702
CAS Number: 35037-73-1
EINECS: 252-328-0
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR