2-Fluorophenyl isothiocyanate ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
A18314.06
A18314.14
A18314.06EA
51.6
EUR
A18314.06
A18314.14
2-Fluorophenyl isothiocyanate ≥97%
2-Fluorophenyl isothiocyanate
Purity: 97%
CAS No.: 38985-64-7
Molecular Formula: C7H4FNS
Formula Weight: 153.18
Flash Point: 97°(206°F)
Refractive Index: 1.6254
Boiling Point: 103-104°/12mm
Beilstein Reference: 4176581
UN No.: 2922
MDL No.: MFCD00004800
Formula:
C₇H₄FNS Boiling Pt: 103…104 °C (12 mmHg) Melting Pt: 24…26 °C Density: 1.248 Flash Pt: 97 °C (206 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004800 CAS Number: 38985-64-7 EINECS: 254-237-1 UN: 2922 ADR: 8,III |