4-(Trifluoromethoxy)aniline, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
337056-25G 337056-5G
SIAL337056-25GEA 71.9 EUR
SIAL337056-25G SIAL337056-5G
4-(Trifluoromethoxy)aniline, Sigma-Aldrich®
4-(Trifluoromethoxy)aniline
Formula: C₇H₆F₃NO
MW: 177.13 g/mol
Boiling Pt: 73
Density: 1.31 g/cm³ (20 °C)
MDL Number: MFCD00041314
CAS Number: 461-82-5
EINECS: 207-317-5
UN: 2810
ADR: 6.1 II